CAS RN 190274-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Calpain Inhibitor VI
The Calpain Inhibitor VI, also referenced under CAS 190274-53-4, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.