accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19047-31-5

IUPAC Name: 2-chloro-N-cyclopropylacetamide
Synonyms: 2-chloro-N-cyclopropylacetamide
Molecular Weight (g/mol): 133.58
Molecular Formula: C5H8ClNO
InChi Key: ZPWIVSGEQGESFF-UHFFFAOYSA-N
SMILES: ClCC(=O)NC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results