accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1906-57-6

IUPAC Name: potassium ethyl oxalate
Synonyms: potassium ethyl oxalate
Molecular Weight (g/mol): 156.18
Molecular Formula: C4H5KO4
InChi Key: RLPQQBNSTHRHEK-UHFFFAOYSA-M
SMILES: [K+].CCOC(=O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)