CAS RN 1906900-37-5
CAS RN 1906900-37-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Calcium Bis(fluorosulfonyl)imide 98.0+%, TCI America™
CAS: 1906900-37-5 Molecular Formula: CaF4N2O8S4