accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CH2C

CAS RN 1907-46-6

IUPAC Name: 3-ethylhex-5-en-3-ol
Synonyms: 3-ethylhex-5-en-3-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: ZNPHYMBSDNDCCM-UHFFFAOYSA-N
SMILES: CCC(O)(CC)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results