accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSOOCH3

CAS RN 1907-65-9

IUPAC Name: N-butyl-4-methylbenzene-1-sulfonamide
Synonyms: N-butyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 227.32
Molecular Formula: C11H17NO2S
InChi Key: RQUXYBHREKXNKT-UHFFFAOYSA-N
SMILES: CCCCNS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results