CAS RN 190788-60-4
IUPAC Name:
2-(2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
234.1
Molecular Formula:
C13H19BO3
InChi Key:
ASDFSWMHZSWXPO-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Sigma Aldrich 2-Methoxyphenylboronic acid, pinacol ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.