CAS RN 19088-73-4
IUPAC Name:
7-hydroxy-2-oxo-2H-chromene-3-carbonitrile
Synonyms:
7-hydroxy-2-oxochromene-3-carbonitrile
Molecular Weight (g/mol):
187.15
Molecular Formula:
C10H5NO3
InChi Key:
IJQYTHQDUDCJEQ-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C=C(C#N)C(=O)OC2=C1
1
–
1
of
1
results
1
3-Cyanoumbelliferone 98.0+%, TCI America™
CAS: 19088-73-4 Molecular Formula: C10H5NO3 Molecular Weight (g/mol): 187.15 MDL Number: MFCD00037480 InChI Key: IJQYTHQDUDCJEQ-UHFFFAOYSA-N Synonym: 3-Cyano-7-hydroxycoumarin PubChem CID: 5393173 IUPAC Name: 7-hydroxy-2-oxo-2H-chromene-3-carbonitrile SMILES: OC1=CC=C2C=C(C#N)C(=O)OC2=C1