accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOSOOOHOONa

CAS RN 19089-60-2

IUPAC Name: sodium 4-carboxy-2-sulfobenzoate
Synonyms: sodium 4-carboxy-2-sulfobenzoate
Molecular Weight (g/mol): 268.17
Molecular Formula: C8H5NaO7S
InChi Key: JARIJYUQOKFVAJ-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)C1=CC(=C(C=C1)C([O-])=O)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results