accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 191-07-1

IUPAC Name: coronene
Synonyms: coronene hexabenzobenzene dibenzo(ghi,pqr)perylene ccris 908
Molecular Weight (g/mol): 300.36
Molecular Formula: C24H12
InChi Key: VPUGDVKSAQVFFS-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C3C=CC4=CC=C5C=CC6=CC=C1C1=C2C3=C4C5=C61
Molecular Weight (g/mol) 
  • (4)
  • (6)
Boiling Point 
  • (7)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (6)
  • (7)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (5)
  • (1)