accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 191-30-0

IUPAC Name: hexacyclo[10.10.2.0²,⁷.0⁸,²⁴.0¹⁵,²³.0¹⁷,²²]tetracosa-1(23),2,4,6,8(24),9,11,13,15,17,19,21-dodecaene
Synonyms: DIBENZO[a,l]PYRENE dibenzo (a,l) pyrene 1,2:3,4-dibenzopyrene DBP DB(a,l)P dibenzo(a,d)pyrene 2,3:4,5-dibenzopyrene 4,5,6,7-dibenzpyrene 1,2,3,4-dibenzopyrene 1,2,9,10-dibenzopyrene
Molecular Weight (g/mol): 302.38
Molecular Formula: C24H14
InChi Key: JNTHRSHGARDABO-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=C3C1=C2C1=CC=CC=C41

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results