accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3CCH3OOCH3NCH3H3CHOCH3OH3CH3CH3CNOONNNH3C

CAS RN 191114-48-4

IUPAC Name: 10-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-4-ethyl-11-methoxy-3a,7,9,11,13,15-hexamethyl-1-{4-[4-(pyridin-3-yl)-1H-imidazol-1-yl]butyl}-tetradecahydro-1H-oxacyclotetradeca[4,3-d][1,3]oxazole-2,6,8,14-tetrone
Synonyms: telithromycin
Molecular Weight (g/mol): 812.02
Molecular Formula: C43H65N5O10
InChi Key: LJVAJPDWBABPEJ-UHFFFAOYNA-N
SMILES: CCC1OC(=O)C(C)C(=O)C(C)C(OC2OC(C)CC(C2O)N(C)C)C(C)(CC(C)C(=O)C(C)C2N(CCCCN3C=NC(=C3)C3=CN=CC=C3)C(=O)OC12C)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results