accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNH3CHOHO

CAS RN 191162-40-0

IUPAC Name: (1-methyl-1H-indol-2-yl)boronic acid
Synonyms: 1-methylindol-2-ylboronic acid
Molecular Weight (g/mol): 174.99
Molecular Formula: C9H10BNO2
InChi Key: CBPBJUTWVXLSER-UHFFFAOYSA-N
SMILES: CN1C(=CC2=CC=CC=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results