accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1912-43-2

IUPAC Name: 2-(2-methyl-1H-indol-3-yl)acetic acid
Synonyms: (2-methyl-1H-indol-3-yl)acetic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: QJNNHJVSQUUHHE-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results