accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OClClCH3HN(S)NH2NSHH

CAS RN 191217-81-9

IUPAC Name: dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride
Synonyms: dihydrogen pramipexole hydrate dichloride
Molecular Weight (g/mol): 302.26
Molecular Formula: C10H21Cl2N3OS
InChi Key: APVQOOKHDZVJEX-QTPLPEIMSA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)