accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3O(E/Z)

CAS RN 19125-76-9

IUPAC Name: 3-(dimethylamino)-2-methylprop-2-enal
Synonyms: 3-(dimethylamino)-2-methylprop-2-enal
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: JGAMOQYFDMQPRJ-UHFFFAOYSA-N
SMILES: CN(C)C=C(C)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results