accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1913-93-5

IUPAC Name: 2-[({3-[1-(3-{[(carboxylatomethyl)(carboxymethyl)azaniumyl]methyl}-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-6-hydroxy-2-methyl-5-(propan-2-yl)phenyl}methyl)(carboxymethyl)azaniumyl]acetate
Synonyms: 2-[({3-[1-(3-{[(carboxylatomethyl)(carboxymethyl)ammonio]methyl}-4-hydroxy-5-isopropyl-2-methylphenyl)-3-oxo-2-benzofuran-1-yl]-6-hydroxy-5-isopropyl-2-methylphenyl}methyl)(carboxymethyl)ammonio]acetate
Molecular Weight (g/mol): 720.77
Molecular Formula: C38H44N2O12
InChi Key: AGXCADFSYJNMEG-UHFFFAOYNA-N
SMILES: CC(C)C1=CC(=C(C)C(C[NH+](CC(O)=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(C[NH+](CC(O)=O)CC([O-])=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results