accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19130-96-2

IUPAC Name: (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
Synonyms: 1-deoxynojirimycin
Molecular Weight (g/mol): 163.17
Molecular Formula: C6H13NO4
InChi Key: LXBIFEVIBLOUGU-JGWLITMVSA-N
SMILES: OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results