accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)OHOHNOOOPOOOH

CAS RN 191348-16-0

IUPAC Name: (2S)-3-(4-{[(benzyloxy)(hydroxy)phosphoryl]oxy}phenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(4-{[benzyloxy(hydroxy)phosphoryl]oxy}phenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 573.54
Molecular Formula: C31H28NO8P
InChi Key: WNFKGEXQEWKECX-KSTXGFNDNA-N
SMILES: OC(=O)[C@H](CC1=CC=C(OP(O)(=O)OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)