CAS RN 19141-85-6
IUPAC Name:
5-ethyl-1H-imidazole
Synonyms:
4-ethyl-3H-imidazole
Molecular Weight (g/mol):
96.13
Molecular Formula:
C5H8N2
InChi Key:
NJQHZENQKNIRSY-UHFFFAOYSA-N
SMILES:
CCC1=CN=CN1
1
–
1
of
1
results
1
4(5)-Ethylimidazole 98.0+%, TCI America™
CAS: 19141-85-6 Molecular Formula: C5H8N2 Molecular Weight (g/mol): 96.133 InChI Key: NJQHZENQKNIRSY-UHFFFAOYSA-N PubChem CID: 10153736 IUPAC Name: 5-ethyl-1H-imidazole SMILES: CCC1=CN=CN1