CAS RN 19145-96-1
IUPAC Name:
1,4-diethyl cyclohexane-1,4-dicarboxylate
Synonyms:
1,4-diethyl cyclohexane-1,4-dicarboxylate
Molecular Weight (g/mol):
228.29
Molecular Formula:
C12H20O4
InChi Key:
KRJHRNUTLDTSKY-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1CCC(CC1)C(=O)OCC
1
–
1
of
1
results
1
Diethyl trans-1,4-Cyclohexanedicarboxylate 98.0+%, TCI America™
CAS: 19145-96-1 Molecular Formula: C12H20O4 Molecular Weight (g/mol): 228.288 InChI Key: KRJHRNUTLDTSKY-UHFFFAOYSA-N Synonym: trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester PubChem CID: 254555 IUPAC Name: diethyl cyclohexane-1,4-dicarboxylate SMILES: CCOC(=O)C1CCC(CC1)C(=O)OCC