accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19158-51-1

IUPAC Name: 4-methylbenzene-1-sulfonyl cyanide
Synonyms: 4-methylbenzenesulfonyl cyanide
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: JONIMGVUGJVFQD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C#N
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results