accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OCH3OH3C

CAS RN 1916-07-0

IUPAC Name: methyl 3,4,5-trimethoxybenzoate
Synonyms: methyl 3,4,5-trimethoxybenzoate
Molecular Weight (g/mol): 226.23
Molecular Formula: C11H14O5
InChi Key: KACHFMOHOPLTNX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(OC)C(OC)=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)