accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnHONHOSSHONOHSS

CAS RN 19163-92-9

IUPAC Name: zinc(2+) bis([bis(2-hydroxyethyl)carbamothioyl]sulfanide)
Synonyms: zinc(2+) bis([bis(2-hydroxyethyl)carbamothioyl]sulfanide)
Molecular Weight (g/mol): 425.9
Molecular Formula: C10H20N2O4S4Zn
InChi Key: BRJVMKALQCUXPN-UHFFFAOYSA-L
SMILES: [Zn++].OCCN(CCO)C([S-])=S.OCCN(CCO)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results