accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClNOHOCl

CAS RN 1918-02-1

IUPAC Name: 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
Synonyms: picloram tordon grazon tordon 22K amdon chloramp k-pin K-pin piclorame tordon 101 mixture
Molecular Weight (g/mol): 241.45
Molecular Formula: C6H3Cl3N2O2
InChi Key: NQQVFXUMIDALNH-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(Cl)=NC(C(O)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results