accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOHOBr

CAS RN 19184-65-7

IUPAC Name: 2-(bromomethyl)-2-(hydroxymethyl)propane-1,3-diol
Synonyms: monobromoneopentyltriol
Molecular Weight (g/mol): 199.04
Molecular Formula: C5H11BrO3
InChi Key: MMHHBAUIJVTLFZ-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)CBr
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)