accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOBr

CAS RN 19199-82-7

IUPAC Name: 4-nitrophenyl 2-bromoacetate
Synonyms: 4-nitrophenyl 2-bromoacetate
Molecular Weight (g/mol): 260.04
Molecular Formula: C8H6BrNO4
InChi Key: HVNXGOPARVAZNX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results