CAS RN 192-65-4
IUPAC Name:
hexacyclo[10.10.2.0²,⁷.0⁸,²⁴.0¹⁵,²³.0¹⁶,²¹]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Synonyms:
dibenzopyrenes DIBENZO[a,e]PYRENE
Molecular Weight (g/mol):
302.38
Molecular Formula:
C24H14
InChi Key:
KGHMWBNEMFNJFZ-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C3=CC=CC=C3C3=CC=CC4=CC=C2C1=C34
1
–
1
of
1
results
1
Dibenzo(a,e)Pyrene, SPEX CertiPrep™
CAS: 192-65-4 Molecular Formula: C24H14 Molecular Weight (g/mol): 302.376 InChI Key: KGHMWBNEMFNJFZ-UHFFFAOYSA-N PubChem CID: 9126 ChEBI: CHEBI:82442 SMILES: C1=CC=C2C3=C4C(=CC2=C1)C5=CC=CC=C5C6=CC=CC(=C64)C=C3