accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONCH3

CAS RN 19205-19-7

IUPAC Name: 5,12-dimethyl-5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
Synonyms: 5,12-dimethyl-5,12-diazapentacene-7,14-dione
Molecular Weight (g/mol): 340.38
Molecular Formula: C22H16N2O2
InChi Key: SCZWJXTUYYSKGF-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2C(=O)C2=CC3=C(C=C12)C(=O)C1=CC=CC=C1N3C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)