accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19214-08-5

IUPAC Name: N-[(2E)-azepan-2-ylidene]hydroxylamine
Synonyms: N-[(2E)-azepan-2-ylidene]hydroxylamine
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: PPOHPUOKXMNCCI-UHFFFAOYSA-N
SMILES: O\N=C1/CCCCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results