accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19221-08-0

IUPAC Name: 4-[2-(4-nitrophenyl)ethenyl]phenol
Synonyms: 4-[2-(4-nitrophenyl)ethenyl]phenol
Molecular Weight (g/mol): 241.25
Molecular Formula: C14H11NO3
InChi Key: OETQWIHJPIESQB-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=CC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results