CAS RN 19227-11-3
IUPAC Name:
N'-hydroxy-2-phenylethanimidamide
Synonyms:
N'-hydroxy-2-phenylethanimidamide
Molecular Weight (g/mol):
150.18
Molecular Formula:
C8H10N2O
InChi Key:
FVYBAJYRRIYNBN-UHFFFAOYSA-N
SMILES:
NC(CC1=CC=CC=C1)=NO
1
–
1
of
1
results
1
N'-Hydroxy-2-phenylethanimidamide ≥97%, Thermo Scientific™
CAS: 19227-11-3 Molecular Formula: C8H10N2O Molecular Weight (g/mol): 150.181 MDL Number: MFCD00019953 InChI Key: FVYBAJYRRIYNBN-UHFFFAOYSA-N Synonym: 2-phenylacetamidoxime,n-hydroxy-2-phenylacetimidamide,1z-n'-hydroxy-2-phenylethanimidamide,n-hydroxy-2-phenyl-acetamidine,z-n'-hydroxy-2-phenylacetimidamide,1-hydroxyimino-2-phenylethylamine,n-hydroxy-2-phenylacetamidine,benzeneethanimidamide,n-hydroxy,benzeneethanimidamide, n-hydroxy,z-n'-hydroxy-2-phenylethanimidamide PubChem CID: 9566465 IUPAC Name: N'-hydroxy-2-phenylethanimidamide SMILES: C1=CC=C(C=C1)CC(=NO)N