accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 192323-07-2

IUPAC Name: tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
Synonyms: tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
Molecular Weight (g/mol): 228.34
Molecular Formula: C12H24N2O2
InChi Key: HMMYZMWDTDJTRR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results