accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNN

CAS RN 19235-89-3

IUPAC Name: 4-chloropyridine-2-carbonitrile
Synonyms: 4-chloropyridine-2-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: DYEZRXLVZMZHQT-UHFFFAOYSA-N
SMILES: ClC1=CC=NC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results