accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19236-15-8

CH3CH3HOCH3CH3HOCH3CH3HONd

CAS RN 19236-15-8

IUPAC Name: tris(propan-2-ol) neodymium
Synonyms: tris(isopropyl alcohol) neodymium
Molecular Weight (g/mol): 324.53
Molecular Formula: C9H24NdO3
InChi Key: WOCLHTSADRFWMF-UHFFFAOYSA-N
SMILES: [Nd].CC(C)O.CC(C)O.CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results