accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrNCS

CAS RN 19241-38-4

IUPAC Name: 4-bromo-1-isothiocyanato-2-methylbenzene
Synonyms: 4-bromo-1-isothiocyanato-2-methylbenzene
Molecular Weight (g/mol): 228.11
Molecular Formula: C8H6BrNS
InChi Key: YASXCQRGYJGIKD-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results