accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19243-95-9

IUPAC Name: N-(4-hydroxyphenyl)-2-methylprop-2-enamide
Synonyms: N-(4-hydroxyphenyl)-2-methylprop-2-enamide
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: XZSZONUJSGDIFI-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)NC1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results