accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1927-06-6

IUPAC Name: (2-hydroxyethyl)trimethylazanium bromide
Synonyms: choline bromide
Molecular Weight (g/mol): 184.08
Molecular Formula: C5H14BrNO
InChi Key: JJCWKVUUIFLXNZ-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)CCO
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)