accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClO

CAS RN 1927-94-2

IUPAC Name: 3-chloro-2-hydroxybenzaldehyde
Synonyms: 3-chloro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: DOHOPUBZLWVZMZ-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC=C1C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results