accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOClClCl

CAS RN 1928-37-6

IUPAC Name: methyl 2-(2,4,5-trichlorophenoxy)acetate
Synonyms: 2,4,5-T, methyl ester
Molecular Weight (g/mol): 269.5
Molecular Formula: C9H7Cl3O3
InChi Key: JUCNGUOYQGHBJC-UHFFFAOYSA-N
SMILES: COC(=O)COC1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results