accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONO

CAS RN 192817-79-1

IUPAC Name: ethyl 2-(morpholin-4-yl)benzoate
Synonyms: ethyl 2-(morpholin-4-yl)benzoate
Molecular Weight (g/mol): 235.28
Molecular Formula: C13H17NO3
InChi Key: UIVDSGDHUXOYDW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results