accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFFFH2NHCl

CAS RN 193140-71-5

IUPAC Name: ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
Synonyms: ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
Molecular Weight (g/mol): 207.58
Molecular Formula: C5H9ClF3NO2
InChi Key: MNFISCBCWBUTTQ-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C(N)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)