accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 193269-78-2

IUPAC Name: tert-butyl 3-aminoazetidine-1-carboxylate
Synonyms: tert-butyl 3-aminoazetidine-1-carboxylate
Molecular Weight (g/mol): 172.23
Molecular Formula: C8H16N2O2
InChi Key: WPGLRFGDZJSQGI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(N)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)