accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 193357-21-0

NHFHCl

CAS RN 193357-21-0

IUPAC Name: 4-[(3-fluorophenyl)methyl]piperidine hydrochloride
Synonyms: 4-[(3-fluorophenyl)methyl]piperidine hydrochloride
Molecular Weight (g/mol): 229.72
Molecular Formula: C12H17ClFN
InChi Key: XZRQMBCQGGXDKQ-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=CC(CC2CCNCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results