CAS RN 193414-60-7
IUPAC Name:
7-bromo-3,4-dihydro-2H-1,4-benzothiazine
Synonyms:
7-bromo-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol):
230.12
Molecular Formula:
C8H8BrNS
InChi Key:
QWWYGAWGUCQKDL-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(NCCS2)C=C1
1
–
1
of
1
results
1
Promotions Available
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
CAS: 193414-60-7 Molecular Formula: C8H8BrNS Molecular Weight (g/mol): 230.12 MDL Number: MFCD11847754 InChI Key: QWWYGAWGUCQKDL-UHFFFAOYSA-N Synonym: 7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine PubChem CID: 83823811 IUPAC Name: 7-bromo-3,4-dihydro-2H-1,4-benzothiazine SMILES: BrC1=CC2=C(NCCS2)C=C1