accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2HNI(E/Z)

CAS RN 193527-91-2

IUPAC Name: N''-(adamantan-2-yl)-N-(4-iodophenyl)guanidine
Synonyms: N''-(adamantan-2-yl)-N-(4-iodophenyl)guanidine
Molecular Weight (g/mol): 395.29
Molecular Formula: C17H22IN3
InChi Key: UUKPIWYXWLJPJF-UHFFFAOYSA-N
SMILES: NC(NC1=CC=C(I)C=C1)=NC1C2CC3CC(C2)CC1C3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results