CAS RN 193693-60-6
CAS RN 193693-60-6
IUPAC Name:
2-[(2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidin-2-yl]acetic acid
Synonyms:
[(2S)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidin-2-yl]acetic acid
Molecular Weight (g/mol):
351.4
Molecular Formula:
C21H21NO4
InChi Key:
ZNIGOUDZWCDFFC-AWEZNQCLSA-N
SMILES:
OC(=O)C[C@@H]1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-Fmoc-L-beta-homoproline, 95%
CAS: 193693-60-6 Molecular Formula: C21H21NO4 Molecular Weight (g/mol): 351.402 MDL Number: MFCD01863058 InChI Key: ZNIGOUDZWCDFFC-AWEZNQCLSA-N Synonym: fmoc-l-beta-homoproline,fmoc-beta-hopro-oh,s-2-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidin-2-yl acetic acid,2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidin-2-yl acetic acid,2-2s-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidin-2-yl acetic acid,fmoc-d-beta-homoproline,fmoc-hopro-oh,fmoc-beta-homopro-oh,fmoc-l-beta-hpro-oh,fmoc-i 2-hopro-oh PubChem CID: 2761542 IUPAC Name: 2-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidin-2-yl]acetic acid SMILES: C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)CC(=O)O