accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOON(S)

CAS RN 193693-66-2

IUPAC Name: (3S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-3-carboxylic acid
Synonyms: (3S)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 337.38
Molecular Formula: C20H19NO4
InChi Key: GUAMYYOQAAUXLR-ZDUSSCGKSA-N
SMILES: OC(=O)[C@H]1CCN(C1)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)