accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNONH2NNNNNH2NH2SOOOSOOONaNa(E/Z)(E/Z)(E/Z)

CAS RN 1937-37-7

IUPAC Name: disodium 5-amino-6-(2-{4'-[2-(2,4-diaminophenyl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium 4-amino-3-(2-{4'-[2-(2,4-diaminophenyl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)-5-oxo-6-(2-phenylhydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 781.73
Molecular Formula: C34H25N9Na2O7S2
InChi Key: OLVNAWLXZDRGPL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=CC(N)=C(C=C1)N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=C(C=C2C=C(C(=NNC3=CC=CC=C3)C(=O)C2=C1N)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)