accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONNHOOOHOHOO

CAS RN 1939-36-2

IUPAC Name: 2-({3-[bis(carboxymethyl)amino]propyl}(carboxymethyl)amino)acetic acid
Synonyms: propanoldiaminetetracetic acid
Molecular Weight (g/mol): 306.27
Molecular Formula: C11H18N2O8
InChi Key: DMQQXDPCRUGSQB-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCCN(CC(O)=O)CC(O)=O)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results